4-Fluoro-[1,1'-biphenyl]-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

48119-1G 48119-25G 48119-500MG 48119-5G
76932-758EA 411.63 USD
76932-758 76932-760 76932-762 76932-764
4-Fluoro-[1,1'-biphenyl]-3-carbaldehyde ≥97%
4-Fluoro-[1,1'-biphenyl]-3-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₉FO
MW: 200.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09042171
CAS Number: 854778-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR