4-Bromo-5-chloro-6-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

47897-1G 47897-25G 47897-500MG 47897-5G
76932-732EA 235.52 USD
76932-732 76932-734 76932-736 76932-738
4-Bromo-5-chloro-6-methyl-1H-indazole ≥97%
4-Bromo-5-chloro-6-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₆BrClN₂
MW: 245.504 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32879462
CAS Number: 2368909-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR