5-Ethoxy-2,4-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

47549-1G 47549-500MG 47549-5G
76932-718EA 558.04 USD
76932-718 76932-720 76932-722
5-Ethoxy-2,4-difluorobenzaldehyde ≥97%
5-Ethoxy-2,4-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD33403862
CAS Number: 2092187-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR