5-Formyl-2,4-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

47276-1G 47276-500MG 47276-5G
76932-704EA 604.72 USD
76932-704 76932-706 76932-708
5-Formyl-2,4-dimethylbenzoic acid ≥97%
5-Formyl-2,4-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD20651233
CAS Number: 1379174-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR