5-Bromo-4-fluoro-2-methoxybenzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

46321-1G 46321-25G 46321-5G
76932-642EA 76.39 USD
76932-642 76932-644 76932-646
5-Bromo-4-fluoro-2-methoxybenzoic acid ≥97% research grade
5-Bromo-4-fluoro-2-methoxybenzoic acid
Formula: C₈H₆BrFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD15527529
CAS Number: 95383-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR