5-Bromo-4-fluoro-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

46265-1G 46265-25G 46265-500MG 46265-5G
76932-626EA 391.48 USD
76932-626 76932-628 76932-630 76932-632
5-Bromo-4-fluoro-2-methoxybenzaldehyde ≥97%
5-Bromo-4-fluoro-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD20923637
CAS Number: 923281-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR