5-Bromo-2-(diethoxymethyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

46017-1G 46017-25G 46017-500MG 46017-5G
76932-570EA 173.99 USD
76932-570 76932-572 76932-574 76932-576
5-Bromo-2-(diethoxymethyl)pyridine ≥97%
5-Bromo-2-(diethoxymethyl)pyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₄BrNO₂
MW: 260.128 g/mol
Storage Temperature: Refrigerator
CAS Number: 1044209-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR