4-Fluoro-3-iodophenol ≥97%

Supplier: AOBChem USA

Warning

45952-1G 45952-25G 45952-500MG 45952-5G
76932-556EA 379.81 USD
76932-556 76932-558 76932-560 76932-562
4-Fluoro-3-iodophenol ≥97%
4-Fluoro-3-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄FIO
MW: 237.998 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17000006
CAS Number: 897956-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR