5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,3-diol ≥97%
Supplier: AOBChem USA
|
Warning
|
45494-1G
45494-25G
45494-500MG
45494-5G
76932-482EA
260.99
USD
76932-482
76932-484
76932-486
76932-488
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,3-diol ≥97%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene-1,3-diol
Precaución: for Research Use Only
Formula:
C₁₂H₁₇BO₄ Storage Temperature: Ambient |
MDL Number:
MFCD22493525 CAS Number: 1352134-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...