5-Bromo-2,3-difluoro-4-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

45125-1G 45125-25G 45125-500MG 45125-5G
76932-466EA 245.07 USD
76932-466 76932-468 76932-470 76932-472
5-Bromo-2,3-difluoro-4-formylphenylboronic acid ≥97%
5-Bromo-2,3-difluoro-4-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₄BBrF₂O₃
MW: 264.817 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30182378
CAS Number: 2377610-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR