4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Carboxy-2-fluorophenylboronic acid pinacol ester

44597-1G 44597-25G 44597-5G
76932-452EA 88.06 USD
76932-452 76932-454 76932-456
4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid ≥97%
4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Formula: C₁₃H₁₆BFO₄
MW: 266.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23098802
CAS Number: 2235384-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR