(4-Chloro-3-formylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-3-formylphenylboronic acid

44360-25G 44360-5G
76932-426EA 268.41 USD
76932-426 76932-428
(4-Chloro-3-formylphenyl)boronic acid ≥97%
(4-Chloro-3-formylphenyl)boronic acid
Formula: C₇H₆BClO₃
MW: 184.38 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28101633
CAS Number: 2354338-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR