4-Carboxy-3-chlorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Carboxy-3-chlorophenylboronic acid, pinacol ester, 2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

41528-1G 41528-25G 41528-5G
76932-416EA 62.6 USD
76932-416 76932-418 76932-420
4-Carboxy-3-chlorophenylboronic acid pinacol ester ≥97%
4-Carboxy-3-chlorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BClO₄
MW: 282.5277 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12546621
CAS Number: 890839-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR