(4-Bromo-2-fluoro-5-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-fluoro-5-methylphenylboronic acid

40329-1G 40329-25G 40329-500MG 40329-5G
76932-388EA 256.74 USD
76932-388 76932-390 76932-392 76932-394
(4-Bromo-2-fluoro-5-methylphenyl)boronic acid ≥97% research grade
(4-Bromo-2-fluoro-5-methylphenyl)boronic acid
Formula: C₇H₇BBrFO₂
Storage Temperature: Ambient
MDL Number: MFCD20441808
CAS Number: 677777-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR