(4-Fluoro-2-methoxy-5-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 4-Fluoro-2-methoxy-5-methylphenylboronic acid

40174-1G 40174-25G 40174-500MG 40174-5G
76932-358EA 221.73 USD
76932-358 76932-360 76932-362 76932-364
(4-Fluoro-2-methoxy-5-methylphenyl)boronic acid ≥97% research grade
(4-Fluoro-2-methoxy-5-methylphenyl)boronic acid
Formula: C₈H₁₀BFO₃
Storage Temperature: Ambient
MDL Number: MFCD18383901
CAS Number: 1207428-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR