5-Benzyloxy-2,4-difluorophenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (5-(Benzyloxy)-2,4-difluorophenyl)boronic acid

40060-1G 40060-25G 40060-5G
76932-346EA 94.43 USD
76932-346 76932-348 76932-350
5-Benzyloxy-2,4-difluorophenylboronic acid ≥95%
5-Benzyloxy-2,4-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BF₂O₃
MW: 264.032 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18783137
CAS Number: 2096335-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR