5-Acetyl-4-bromothiophen-2-boronic acid ≥97%

Supplier: AOBChem USA

Warning

39777-1G 39777-25G 39777-500MG 39777-5G
76932-318EA 245.07 USD
76932-318 76932-320 76932-322 76932-324
5-Acetyl-4-bromothiophen-2-boronic acid ≥97%
5-Acetyl-4-bromothiophen-2-boronic acid

Precaución: for Research Use Only

Formula: C₆H₆BBrO₃S
Storage Temperature: Refrigerator
MDL Number: MFCD16660286
CAS Number: 1256346-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR