2-Chloro-5-bromo-4-(trifluoromethyl)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-chloro-4-(trifluoromethyl)phenylboronic acid, (5-Bromo-2-chloro-4-(trifluoromethyl)phenyl)boronic acid

39247-1G 39247-25G 39247-5G
76932-304EA 114.58 USD
76932-304 76932-306 76932-308
2-Chloro-5-bromo-4-(trifluoromethyl)phenylboronic acid ≥97% research grade
2-Chloro-5-bromo-4-(trifluoromethyl)phenylboronic acid
Formula: C₇H₄BBrClF₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD12026719
CAS Number: 1452574-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR