4-Bromo-5-chloro-2-methylbenzonitrile ≥95%
Supplier: AOBChem USA
|
Warning
|
38741-1G
38741-500MG
38741-5G
76932-294EA
524.09
USD
76932-294
76932-296
76932-298
4-Bromo-5-chloro-2-methylbenzonitrile ≥95%
4-Bromo-5-chloro-2-methylbenzonitrile
Precaución: for Research Use Only
Formula:
C₈H₅BrClN MW: 230.489 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18917160 CAS Number: 1228376-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...