5-Bromo-4-chloro-3-iodo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

38707-1G 38707-25G 38707-5G
76932-268EA 88.06 USD
76932-268 76932-270 76932-272
5-Bromo-4-chloro-3-iodo-1H-indazole ≥97%
5-Bromo-4-chloro-3-iodo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₃BrClIN₂
MW: 357.374 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11007985
CAS Number: 1082040-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR