4-Bromo-5-methyl-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole ≥95% research grade

Supplier: AOBChem USA

Warning

38574-1G 38574-25G 38574-5G
76932-258EA 109.28 USD
76932-258 76932-260 76932-262
4-Bromo-5-methyl-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole ≥95% research grade
4-Bromo-5-methyl-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole
Formula: C₁₃H₁₅BrN₂O
MW: 295.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19443358
CAS Number: 1926172-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR