2,3-Dimethyl-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Methoxy-2,3-dimethylbenzoic acid

38494-1G 38494-25G 38494-500MG 38494-5G
76932-242EA 284.33 USD
76932-242 76932-244 76932-246 76932-248
2,3-Dimethyl-4-methoxybenzoic acid ≥97%
2,3-Dimethyl-4-methoxybenzoic acid
Formula: C₁₀H₁₂O₃
MW: 180.21 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06203271
CAS Number: 5628-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR