4-Chloro-2,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

38404-1G 38404-25G 38404-500MG 38404-5G
76932-228EA 266.29 USD
76932-228 76932-230 76932-232 76932-234
4-Chloro-2,5-dimethylbenzoic acid ≥97%
4-Chloro-2,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11845877
CAS Number: 90649-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR