4-Ethoxy-2,5-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

38334-1G 38334-500MG 38334-5G
76932-214EA 706.56 USD
76932-214 76932-216 76932-218
4-Ethoxy-2,5-difluorobenzaldehyde ≥97%
4-Ethoxy-2,5-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06247324
CAS Number: 883535-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR