4-Bromo-6-fluoro-7-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

38133-1G 38133-500MG 38133-5G
76932-188EA 468.92 USD
76932-188 76932-190 76932-192
4-Bromo-6-fluoro-7-methyl-1H-indazole ≥97%
4-Bromo-6-fluoro-7-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₆BrFN₂
Storage Temperature: Refrigerator
MDL Number: MFCD32632307
CAS Number: 2404734-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR