4-Bromo-2-fluoro-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

38100-1G 38100-25G 38100-5G
76932-176EA 108.22 USD
76932-176 76932-178 76932-180
4-Bromo-2-fluoro-3-methylbenzoic acid ≥97%
4-Bromo-2-fluoro-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22052783
CAS Number: 194804-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR