2-Amino-5-(p-tolyl)thiazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-(p-Tolyl)thiazol-2-amine

37975-1G 37975-25G 37975-500MG 37975-5G
76932-150EA 154.9 USD
76932-150 76932-152 76932-154 76932-156
2-Amino-5-(p-tolyl)thiazole ≥97%
2-Amino-5-(p-tolyl)thiazole

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂S
MW: 190.265 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00858269
CAS Number: 73040-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR