5-Fluoro-4-methoxy-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37867-1G 37867-25G 37867-500MG 37867-5G
76932-134EA 439.22 USD
76932-134 76932-136 76932-138 76932-140
5-Fluoro-4-methoxy-2-methylbenzoic acid ≥97%
5-Fluoro-4-methoxy-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22414694
CAS Number: 1427448-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR