4-methoxy-1H-indazol-3-amine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Amino-4-methoxy-1H-indazole, 4-Methoxy-1H-indazol-3-amine

37836-1G 37836-25G 37836-5G
76932-120EA 62.6 USD
76932-120 76932-122 76932-124
4-methoxy-1H-indazol-3-amine ≥97%
4-methoxy-1H-indazol-3-amine
Formula: C₈H₉N₃O
MW: 163.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08276946
CAS Number: 886362-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR