2-(4-Fluorophenyl)benzonitrile ≥97% research grade

Supplier: AOBChem USA

Warning

37773-25G 37773-5G
76932-108EA 595.17 USD
76932-108 76932-110
2-(4-Fluorophenyl)benzonitrile ≥97% research grade
2-(4-Fluorophenyl)benzonitrile
Formula: C₁₃H₈FN
Storage Temperature: Ambient
MDL Number: MFCD05979720
CAS Number: 89346-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR