4-Ethoxy-2-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37726-1G 37726-25G 37726-500MG 37726-5G
76932-092EA 406.33 USD
76932-092 76932-094 76932-096 76932-098
4-Ethoxy-2-formylbenzoic acid ≥97%
4-Ethoxy-2-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564052
CAS Number: 2734774-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR