5-Bromo-2,3-difluoro-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37439-1G 37439-25G 37439-500MG 37439-5G
76932-044EA 244.01 USD
76932-044 76932-046 76932-048 76932-050
5-Bromo-2,3-difluoro-4-methoxybenzoic acid ≥97%
5-Bromo-2,3-difluoro-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD32632219
CAS Number: 1780570-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR