4-Hydroxy-3,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

37415-1G 37415-500MG 37415-5G
76932-032EA 589.87 USD
76932-032 76932-034 76932-036
4-Hydroxy-3,5-dimethylbenzoic acid ≥97%
4-Hydroxy-3,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00016536
CAS Number: 4919-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR