5-Bromo-2-butoxy-1,3-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

37377-25G 37377-5G
76932-022EA 453.01 USD
76932-022 76932-024
5-Bromo-2-butoxy-1,3-dimethylbenzene ≥97%
5-Bromo-2-butoxy-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₇BrO
Storage Temperature: Ambient
MDL Number: MFCD18917838
CAS Number: 370104-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR