4-Bromo-2-fluoro-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

37253-1G 37253-25G 37253-5G
76931-992EA 62.6 USD
76931-992 76931-994 76931-996
4-Bromo-2-fluoro-3-methoxybenzaldehyde ≥97%
4-Bromo-2-fluoro-3-methoxybenzaldehyde
Formula: C₈H₆BrFO₂
MW: 233.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27578163
CAS Number: 1820614-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR