4-Methoxy-4'-nitrobiphenyl ≥95%

Supplier: AOBChem USA

Warning

37108-1G 37108-25G 37108-500MG 37108-5G
76931-976EA 138.98 USD
76931-976 76931-978 76931-980 76931-982
4-Methoxy-4'-nitrobiphenyl ≥95%
4-Methoxy-4'-nitrobiphenyl
Formula: C₁₃H₁₁NO₃
Storage Temperature: Ambient
MDL Number: MFCD00143334
CAS Number: 2143-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR