4-Chloro-3-fluoro-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

36911-1G 36911-500MG 36911-5G
76931-950EA 554.86 USD
76931-950 76931-952 76931-954
4-Chloro-3-fluoro-2-methoxybenzaldehyde ≥97%
4-Chloro-3-fluoro-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
MW: 188.583 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24686301
CAS Number: 1554658-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR