5-Chloro-2-fluorophenetole ≥97%

Supplier: AOBChem USA

Warning

36792-25G 36792-5G
76931-942EA 346.92 USD
76931-942 76931-944
5-Chloro-2-fluorophenetole ≥97%
5-Chloro-2-fluorophenetole
Formula: C₈H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD00672980
CAS Number: 289039-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR