5-Chloro-2-fluoroanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Chloro-1-fluoro-2-methoxybenzene

36788-25G 36788-5G
76931-930EA 240.83 USD
76931-930 76931-932
5-Chloro-2-fluoroanisole ≥97%
5-Chloro-2-fluoroanisole

Precaución: for Research Use Only

Formula: C₇H₆ClFO
MW: 160.58 g/mol
Storage Temperature: Ambient
CAS Number: 1092349-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR