4'-Methoxy-2-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

36784-1G 36784-25G 36784-500MG 36784-5G
76931-914EA 326.76 USD
76931-914 76931-916 76931-918 76931-920
4'-Methoxy-2-methyl-1,1'-biphenyl ≥97%
4'-Methoxy-2-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₄O
Storage Temperature: Refrigerator
MDL Number: MFCD06802485
CAS Number: 92495-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR