4-Fluoro-2-isopropoxy-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

36605-25G 36605-5G
76931-878EA 630.18 USD
76931-878 76931-880
4-Fluoro-2-isopropoxy-1-methylbenzene ≥97%
4-Fluoro-2-isopropoxy-1-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO
MW: 168.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23113737
CAS Number: 1369835-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR