4-Chloro-1,2-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

36533-25G 36533-5G
76931-862EA 407.39 USD
76931-862 76931-864
4-Chloro-1,2-dimethoxybenzene ≥95%
4-Chloro-1,2-dimethoxybenzene
Formula: C₈H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD01741851
CAS Number: 16766-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR