5-(Ethoxycarbonyl)-2-fluorophenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (5-(Ethoxycarbonyl)-2-fluorophenyl)boronic acid, 5-Ethoxycarbonyl-2-fluorobenzeneboronic acid

36419-25G 36419-5G
76931-834EA 445.58 USD
76931-834 76931-836
5-(Ethoxycarbonyl)-2-fluorophenylboronic acid ≥95%
5-(Ethoxycarbonyl)-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BFO₄
Storage Temperature: Ambient
MDL Number: MFCD08235094
CAS Number: 874219-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR