4-Methoxy-3-(methoxymethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

36361-1G 36361-500MG 36361-5G
76931-820EA 587.74 USD
76931-820 76931-822 76931-824
4-Methoxy-3-(methoxymethyl)benzoic acid ≥97%
4-Methoxy-3-(methoxymethyl)benzoic acid
Formula: C₁₀H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD02856325
CAS Number: 91061-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR