4-Bromo-N,3-dimethyl-N-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-N-methoxy-N,3-dimethylbenzamide

35854-1G 35854-25G 35854-500MG 35854-5G
76931-772EA 135.8 USD
76931-772 76931-774 76931-776 76931-778
4-Bromo-N,3-dimethyl-N-methoxybenzamide ≥97%
4-Bromo-N,3-dimethyl-N-methoxybenzamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂BrNO₂
MW: 258.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26802226
CAS Number: 170230-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR