5-(4-Methoxyphenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35823-1G 35823-25G 35823-500MG 35823-5G
76931-756EA 126.25 USD
76931-756 76931-758 76931-760 76931-762
5-(4-Methoxyphenyl)oxazole ≥97%
5-(4-Methoxyphenyl)oxazole

Precaución: for Research Use Only

Formula: C₁₀H₉NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00085108
CAS Number: 1011-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR