5-(3-Fluorophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35821-1G 35821-25G 35821-500MG 35821-5G
76931-740EA 260.99 USD
76931-740 76931-742 76931-744 76931-746
5-(3-Fluorophenyl)oxazole ≥97%
5-(3-Fluorophenyl)oxazole
Formula: C₉H₆FNO
MW: 163.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12024926
CAS Number: 243455-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR