5-Acetyl-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

35811-1G 35811-25G 35811-500MG 35811-5G
76931-726EA 269.47 USD
76931-726 76931-728 76931-730 76931-732
5-Acetyl-2-methoxybenzoic acid ≥97%
5-Acetyl-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
Storage Temperature: Refrigerator
MDL Number: MFCD12827984
CAS Number: 68535-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR