5-(2-Bromophenyl)oxazole ≥97%

Supplier: AOBChem USA

Warning

35736-25G 35736-5G
76931-708EA 289.63 USD
76931-708 76931-710
5-(2-Bromophenyl)oxazole ≥97%
5-(2-Bromophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD05668985
CAS Number: 328270-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR