5-Bromo-2-fluoro-3-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

35401-1G 35401-25G 35401-5G
76931-654EA 96.55 USD
76931-654 76931-656 76931-658
5-Bromo-2-fluoro-3-methoxyphenylboronic acid ≥97%
5-Bromo-2-fluoro-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD22421497
CAS Number: 2377610-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR